Assay Details
Target Gene Details
Entrez Gene ID: | 11801 |
Gene Name: | CD5 antigen-like |
Gene Aliases: |
1/6, AAC-11, AI047839, Api6, CT2, Pdp, Sp-alpha |
Location: |
Chr.3:87357881-87371074 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd5l | NM_009690.2 | 5 | 1001 | NP_033820.2 |
| AF011428.1 | 5 | 907 | AAD01445.1 | |
| AF018268.1 | 5 | 940 | AAB70571.1 | |
| AF018269.1 | 5 | 926 | AAB70572.1 | |
| AK159132.1 | 5 | 950 | BAE34844.1 | |
| AK159181.1 | 5 | 1004 | BAE34880.1 | |
| AK159823.1 | 5 | 969 | BAE35403.1 | |
| BC006799.1 | 5 | 942 | AAH06799.1 | |
| BC094459.1 | 5 | 954 | AAH94459.1 |

Genomic Map