Assay Details
Target Gene Details
Entrez Gene ID: | 57912 |
Gene Name: | CDC42 small effector 1 |
Gene Aliases: |
1300002M12Rik, AW558204, Cdcse1, SCIP1, Spec1 |
Location: |
Chr.3:95228424-95236424 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdc42se1 | NM_001038708.3 | 5 | 637 | NP_001033797.1 |
| NM_172395.3 | 5 | 1007 | NP_765983.1 | |
| AK004859.1 | 5 | 652 | BAC25099.1 | |
| AK008084.1 | 4 | 678 | ||
| AK019052.1 | 5 | 649 | BAC25576.1 | |
| AK028573.1 | 5 | 649 | BAC26014.1 | |
| AK213146.1 | 3 | 223 | ||
| BC060964.1 | 5 | 629 | AAH60964.1 |

Genomic Map