Assay Details
Target Gene Details
Entrez Gene ID: | 13559 |
Gene Name: | E2F transcription factor 5 |
Gene Aliases: |
AU024671, E2F-5 |
Location: |
Chr.3:14578602-14606309 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| E2f5 | NM_007892.2 | 8 | 1622 | NP_031918.2 |
| AK156760.1 | 8 | 1541 | BAE33842.1 | |
| AK191418.1 | 1 | 130 | ||
| BC003220.1 | 8 | 1622 | AAH03220.1 |
More Information
Set Membership: |
|

Genomic Map