Assay Details
Target Gene Details
Entrez Gene ID: | 20194 |
Gene Name: | S100 calcium binding protein A10 (calpactin) |
Gene Aliases: |
42C, AA409961, AL024248, CAL12, CLP11, Cal1l, p10, p11 |
Location: |
Chr.3:93555117-93564645 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100a10 | NM_009112.2 | 3 | 519 | NP_033138.1 |
| AK149077.1 | 3 | 505 | BAE28731.1 | |
| AK161647.1 | 3 | 503 | BAE36511.1 | |
| AK170919.1 | 3 | 499 | BAE42112.1 | |
| BC025044.1 | 3 | 450 | AAH25044.1 | |
| D18288.1 | ||||
| M16465.1 | 3 | 472 | AAA37363.1 |
More Information
Set Membership: |
|

Genomic Map