Assay Details
Target Gene Details
Entrez Gene ID: | 99681 |
Gene Name: | trichohyalin |
Gene Aliases: |
AHF, AI597080, Thh |
Location: |
Chr.3:93442330-93449077 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tchh | NM_001163098.1 | 2 | 2038 | NP_001156570.1 |
| AK132147.1 | 1 | 646 | BAE20996.1 |

Genomic Map