Assay Details
Target Gene Details
Entrez Gene ID: | 229096 |
Gene Name: | YTH domain family 3 |
Gene Aliases: |
9130022A11Rik |
Location: |
Chr.3:16183005-16217037 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ythdf3 | NM_001145919.1 | 4 | 1778 | NP_001139391.1 |
| NM_172677.3 | 5 | 1811 | NP_766265.3 | |
| NR_027375.1 | 5 | 1927 | ||
| AK039176.1 | 5 | 1874 | BAC30267.1 | |
| AK053736.1 | 5 | 1846 | BAC35498.1 | |
| AK078922.1 | 1 | 546 | BAC37461.1 | |
| AK137245.1 | 5 | 1758 | BAE23284.1 | |
| BC022932.1 | 1 | 236 | AAH22932.1 | |
| BC052631.1 | 1 | 1128 | AAH52631.1 | |
| BC057158.1 | 5 | 1838 | AAH57158.1 | |
| BC067040.1 | 4 | 1749 | AAH67040.1 | |
| BC067042.1 | 5 | 1744 | AAH67042.1 |

Genomic Map