Assay Details
Target Gene Details
Entrez Gene ID: | 229096 |
Gene Name: | YTH domain family 3 |
Gene Aliases: |
9130022A11Rik |
Location: |
Chr.3:16183005-16217037 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ythdf3 | NM_001145919.1 | 5 | 4096 | NP_001139391.1 |
| NM_172677.3 | 6 | 4129 | NP_766265.3 | |
| NR_027375.1 | 6 | 4245 | ||
| BC067040.1 | 5 | 4067 | AAH67040.1 | |
| BC067042.1 | 6 | 4062 | AAH67042.1 |

Genomic Map