Assay Details
Target Gene Details
Entrez Gene ID: | 140475 |
Gene Name: | Bartter syndrome, infantile, with sensorineural deafness (Barttin) |
Gene Aliases: |
- |
Location: |
Chr.4:106483456-106492243 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bsnd | NM_080458.2 | 4 | 1724 | NP_536706.2 |
| AF391088.1 | AAL33907.1 | |||
| AK052587.1 | 4 | 1725 | BAC35049.1 | |
| BC038287.1 | 4 | 1775 | AAH38287.1 |

Genomic Map