Assay Details
Target Gene Details
Entrez Gene ID: | 18598 |
Gene Name: | pyruvate dehydrogenase E1 alpha 2 |
Gene Aliases: |
Pdhal |
Location: |
Chr.3:141210004-141212355 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdha2 | NM_008811.2 | 1 | 1173 | NP_032837.1 |
| AK006356.1 | 1 | 602 | BAB24543.1 | |
| AK076791.1 | 1 | 645 | BAC36482.1 | |
| BC100460.1 | 1 | 593 | AAI00461.1 | |
| M76728.1 | 1 | 1171 | AAA53047.1 |
More Information
Set Membership: |
|

Genomic Map