Assay Details
Target Gene Details
Entrez Gene ID: | 20200 |
Gene Name: | S100 calcium binding protein A6 (calcyclin) |
Gene Aliases: |
2A9, 5B10, CALCYCLIN, Cacy, PRA |
Location: |
Chr.3:90612894-90614414 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100a6 | NM_011313.2 | 1 | 112 | NP_035443.1 |
| BM932500.1 | 1 | 124 |
More Information
Set Membership: |
|

Genomic Map