Assay Details
Target Gene Details
Entrez Gene ID: | 75015 |
Gene Name: | RIKEN cDNA 4930503B20 gene |
Gene Aliases: |
1700013A07Rik, 1700095G21Rik, 4930483N21Rik, Gm1652, Samd13 |
Location: |
Chr.3:146645189-146686801 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930503B20Rik |

Genomic Map