Assay Details
Target Gene Details
Entrez Gene ID: | 80892 |
Gene Name: | zinc finger homeodomain 4 |
Gene Aliases: |
A930021B15, C130041O22Rik, Zfh-4, Zfh4 |
Location: |
Chr.3:5217420-5415861 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfhx4 | NM_030708.2 | 2 | 1688 | NP_109633.2 |
| AB024499.1 | 1 | 1312 | BAB03600.1 | |
| AK044540.1 | 2 | 1584 | BAC31970.1 | |
| AK084213.1 |
More Information
Set Membership: |
|

Genomic Map