Assay Details
Target Gene Details
Entrez Gene ID: | 73353 |
Gene Name: | actin-related protein T2 |
Gene Aliases: |
1700052K15Rik, Arp-T2, Arpm2 |
Location: |
Chr.4:154666428-154667867 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Actrt2 | NM_028513.3 | 1 | 481 | NP_082789.1 |
| AK006772.1 | 1 | 326 | BAB24733.1 | |
| AK077010.2 | 1 | 482 | ||
| BC052344.1 | 1 | 347 | AAH52344.1 | |
| BU946350.1 | 1 | 483 |
More Information
Set Membership: |
|

Genomic Map