Assay Details
Target Gene Details
Entrez Gene ID: | 70533 |
Gene Name: | basic transcription factor 3-like 4 |
Gene Aliases: |
4632412E09Rik, 5730434I03Rik, AI316863, AU022207, AW552446 |
Location: |
Chr.4:108814295-108834297 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Btf3l4 | NM_027453.2 | 6 | 2689 | NP_081729.1 |
| AK076283.1 | 6 | 2689 | BAC36287.1 |
More Information
Set Membership: |
|

Genomic Map