Assay Details
Target Gene Details
Entrez Gene ID: | 12870 |
Gene Name: | ceruloplasmin |
Gene Aliases: |
D3Ertd555e |
Location: |
Chr.3:19956987-20009750 on Build GRCm38 |
Assay Gene Location: | Within Intron 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cp | NM_001276248.1 | NP_001263177.1 | ||
| NM_001276250.1 | NP_001263179.1 | |||
| AK014835.1 | ||||
| AK043248.1 | ||||
| AK086999.1 | BAC39781.1 |
Target Gene Details
Entrez Gene ID: | 12807 |
Gene Name: | Hermansky-Pudlak syndrome 3 homolog (human) |
Gene Aliases: |
coa |
Location: |
Chr.3:19995945-20035376 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hps3 | NM_001146323.1 | 17 | 2715 | NP_001139795.1 |
| NM_080634.4 | 17 | 3111 | NP_542365.3 | |
| AF393747.1 | 17 | 3100 | AAK84181.1 | |
| AF393780.1 | 17 | 3103 | AAL84109.1 | |
| AK034377.1 | 17 | 3082 | BAC28690.1 | |
| AK036663.1 | 17 | 3119 | BAC29524.1 | |
| AK155497.1 | 17 | 3079 | ||
| AK157953.1 | 17 | 3097 | ||
| AK169855.1 | 16 | 3183 | ||
| AK170581.1 | 17 | 2627 | BAE41891.1 | |
| AK171997.1 | 17 | 2703 | BAE42766.1 | |
| BC020105.1 | 7 | 1210 | AAH20105.1 | |
| BC058235.1 | 17 | 3085 | AAH58235.1 |
More Information
Set Membership: |
|

Genomic Map