Assay Details
Target Gene Details
Entrez Gene ID: | 67161 |
Gene Name: | sodium channel and clathrin linker 1 |
Gene Aliases: |
2610207F23Rik, 4931421F20Rik |
Location: |
Chr.3:41626705-41742530 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sclt1 | NM_001081411.1 | 21 | 2664 | NP_001074880.1 |
| AK089007.1 | 10 | 1156 | ||
| BC043136.1 | 21 | 2646 | AAH43136.1 | |
| BC060271.1 | 21 | 2646 | AAH60271.1 |

Genomic Map