Assay Details
Target Gene Details
Entrez Gene ID: | 11444 |
Gene Name: | cholinergic receptor, nicotinic, beta polypeptide 2 (neuronal) |
Gene Aliases: |
Acrb-2, Acrb2, C030030P04Rik, [b]2-nAchR |
Location: |
Chr.3:89753448-89764632 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chrnb2 | NM_009602.4 | 6 | 3479 | NP_033732.2 |
| AK049722.1 | 6 | 3452 | BAC33893.1 | |
| AK051742.1 | 6 | 3461 | BAC34749.1 | |
| BC065103.1 | 6 | 3432 | AAH65103.1 |
More Information
Set Membership: |
|

Genomic Map