Assay Details
Target Gene Details
Entrez Gene ID: | 67306 |
Gene Name: | zinc finger, C2HC-type containing 1A |
Gene Aliases: |
3110050N22Rik, AI790358, AU023959, Fam164a |
Location: |
Chr.3:7503415-7553848 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zc2hc1a | NM_173181.3 | 9 | 2534 | NP_775273.1 |
| AK012273.1 | 1 | 1349 | ||
| AK034783.1 | 2 | 1680 | ||
| AK083941.1 | 9 | 2478 | BAC39075.1 | |
| AK085187.1 | 9 | 2472 |
More Information
Set Membership: |
|

Genomic Map