Assay Details
Target Gene Details
Entrez Gene ID: | 15496 |
Gene Name: | hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 5 |
Gene Aliases: |
- |
Location: |
Chr.3:98618636-98645534 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hsd3b5 | NM_008295.2 | 4 | 1412 | NP_032321.2 |
| BC012715.1 | 4 | 1416 | AAH12715.1 | |
| BY460328.1 | 1 | 272 | ||
| L41519.1 | 4 | 1397 | AAB81242.1 |
More Information
Set Membership: |
|

Genomic Map