Assay Details
Target Gene Details
Entrez Gene ID: | 75624 |
Gene Name: | methionyl aminopeptidase 1 |
Gene Aliases: |
1700029C17Rik, AW047992, mKIAA0094 |
Location: |
Chr.3:138458960-138489382 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Metap1 | NM_175224.4 | 11 | 1775 | NP_780433.1 |
| AK077694.1 | 11 | 1776 | BAC36961.1 | |
| AK129054.1 | 6 | 3882 | BAC97864.2 | |
| AK153944.1 | 11 | 1782 | ||
| BC027110.1 | 6 | 1315 | AAH27110.1 | |
| BC096469.1 | 11 | 1766 | AAH96469.1 |
More Information
Set Membership: |
|

Genomic Map