Assay Details
Target Gene Details
Entrez Gene ID: | 57912 |
Gene Name: | CDC42 small effector 1 |
Gene Aliases: |
1300002M12Rik, AW558204, Cdcse1, SCIP1, Spec1 |
Location: |
Chr.3:95228424-95236424 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdc42se1 | NM_172395.3 | 1 | 149 | NP_765983.1 |
| AV456394.1 | 1 | 149 |
Target Gene Details
Entrez Gene ID: | 56772 |
Gene Name: | myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 11 |
Gene Aliases: |
AI839562, Af1q, Zfp692 |
Location: |
Chr.3:95218544-95230459 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mllt11 | NM_019914.4 | 1 | 106 | NP_064298.1 |
| AK152708.1 | 1 | 106 | BAE31435.1 |

Genomic Map