Assay Details
Target Gene Details
Entrez Gene ID: | 100504029 |
Gene Name: | predicted gene, 16740 |
Gene Aliases: |
- |
Location: |
Chr.3:95217457-95226658 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm16740 | NR_108026.1 | |||
| AK020406.1 |
Target Gene Details
Entrez Gene ID: | 56772 |
Gene Name: | myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 11 |
Gene Aliases: |
AI839562, Af1q, Zfp692 |
Location: |
Chr.3:95218544-95230459 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mllt11 | NM_019914.4 | 2 | 1233 | NP_064298.1 |
| AB000733.1 | 2 | 807 | BAA19173.1 | |
| AK046005.1 | 2 | 738 | BAC32568.1 | |
| AK152708.1 | 2 | 1233 | BAE31435.1 | |
| AK158495.1 | 2 | 859 | BAE34537.1 | |
| BC022963.1 | 2 | 674 | AAH22963.1 | |
| U95498.1 | 2 | 794 | AAD00803.1 |
More Information
Set Membership: |
|

Genomic Map