Assay Details
Target Gene Details
Entrez Gene ID: | 229302 |
Gene Name: | transmembrane 4 superfamily member 4 |
Gene Aliases: |
Iltmp |
Location: |
Chr.3:57425410-57441677 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tm4sf4 | NM_145539.2 | 5 | 1291 | NP_663514.2 |
| AK148484.1 | 5 | 1259 | BAE28579.1 | |
| AK165731.1 | BAE38358.1 | |||
| BC010814.1 | 5 | 1242 | AAH10814.1 | |
| BP772386.1 | 1 | 153 |

Genomic Map