Assay Details
Target Gene Details
Entrez Gene ID: | 71913 |
Gene Name: | transmembrane protein 79 |
Gene Aliases: |
2310042N02Rik, 2310074C17Rik |
Location: |
Chr.3:88328653-88334433 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem79 | NM_024246.5 | 4 | 1907 | NP_077208.1 |
| AK009760.1 | 4 | 1908 | BAB26484.1 | |
| AK010144.1 | ||||
| BC003309.1 | AAH03309.1 |

Genomic Map