Assay Details
Target Gene Details
Entrez Gene ID: | 69769 |
Gene Name: | tumor necrosis factor, alpha-induced protein 8-like 2 |
Gene Aliases: |
1810019A08Rik, AW610835, TIPE2 |
Location: |
Chr.3:95139521-95142448 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnfaip8l2 | NM_027206.2 | 2 | 402 | NP_081482.1 |
| AF548004.2 | 1 | 310 | ||
| AK007540.1 | 2 | 403 | BAB25098.1 | |
| BC055879.1 | 2 | 377 | AAH55879.1 | |
| BC094616.1 | 2 | 406 | AAH94616.1 |

Genomic Map