Assay Details
Target Gene Details
Entrez Gene ID: | 73721 |
Gene Name: | RIKEN cDNA 1110017D15 gene |
Gene Aliases: |
Cbe1, Smrp1 |
Location: |
Chr.4:41505005-41517333 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1110017D15Rik | NM_001048005.1 | 1 | 154 | NP_001041470.1 |
| NM_001253777.1 | 1 | 154 | NP_001240706.1 | |
| NM_001253788.1 | 1 | 154 | NP_001240717.1 | |
| NR_045549.2 | 1 | 154 | ||
| AB231294.1 | 1 | 154 | BAE78528.1 |

Genomic Map