Assay Details
Target Gene Details
Entrez Gene ID: | 71861 |
Gene Name: | zinc finger SWIM-type containing 2 |
Gene Aliases: |
1700025P14Rik, 4933437F18Rik, MEX |
Location: |
Chr.2:83915079-83941254 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zswim2 | NM_027964.2 | 9 | 1613 | NP_082240.1 |
| AK006367.1 | 9 | 1614 | BAB24549.1 | |
| BC120837.1 | 9 | 1593 | ||
| EU233996.1 | 7 | 1728 |
More Information
Set Membership: |
|

Genomic Map