Assay Details
Target Gene Details
Entrez Gene ID: | 67538 |
Gene Name: | zinc finger SWIM-type containing 3 |
Gene Aliases: |
4921517A06Rik, C86566 |
Location: |
Chr.2:164805114-164822127 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zswim3 | NM_178375.2 | 2 | 1112 | NP_848462.1 |
| AK014904.1 | 2 | 1132 | ||
| AK016241.1 | 1 | 586 | ||
| BC023287.1 | 2 | 1112 | AAH23287.1 |

Genomic Map