Assay Details
Target Gene Details
Entrez Gene ID: | 99696 |
Gene Name: | ankyrin repeat domain 50 |
Gene Aliases: |
AI662170, E430012K20Rik, mKIAA1223, mKIAA3019 |
Location: |
Chr.3:38449261-38484861 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankrd50 | NM_001167883.1 | 4 | 1958 | NP_001161355.1 |
| AK129484.1 | 4 | 1943 | BAC98294.1 | |
| AK147106.1 | 2 | 2098 | BAE27678.1 | |
| AK162481.1 | 2 | 1806 | BAE36940.1 | |
| AK220375.1 | 2 | 2095 | BAD90249.1 | |
| BC145902.1 |
More Information
Set Membership: |
|

Genomic Map