Assay Details
Target Gene Details
Entrez Gene ID: | 99899 |
Gene Name: | interferon-induced protein 44 |
Gene Aliases: |
A430056A10Rik, AW261460, MTAP44, p44 |
Location: |
Chr.3:151730922-151749959 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ifi44 | NM_133871.2 | 8 | 1549 | NP_598632.2 |
| AJ299405.1 | 7 | 1439 | CAC13979.1 | |
| AK079749.1 | 8 | 1501 | BAC37740.1 | |
| AK136553.1 | 8 | 1501 | BAE23041.1 | |
| AK151346.1 | 8 | 1500 | ||
| AK171676.1 | 8 | 1482 | BAE42605.1 | |
| BC026901.1 | 3 | 447 | AAH26901.1 | |
| BC111882.1 | 7 | 1439 |

Genomic Map