Assay Details
Target Gene Details
Entrez Gene ID: | 433693 |
Gene Name: | akirin 2 |
Gene Aliases: |
2700059D21Rik, AA114675, AA522011, AU019887, Akirin-2 |
Location: |
Chr.4:34550615-34566911 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Akirin2 | NM_001007589.3 | 5 | 1256 | NP_001007590.2 |
| AK004133.1 | 1 | 201 | ||
| AK005836.1 | 5 | 938 | BAB24265.1 |

Genomic Map