Assay Details
Target Gene Details
Entrez Gene ID: | 68659 |
Gene Name: | family with sequence similarity 198, member B |
Gene Aliases: |
1110032E23Rik, 2210419I08Rik, AV011458, Ened |
Location: |
Chr.3:79884475-79946278 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam198b | NM_133187.3 | 4 | 4954 | NP_573450.2 |
| AK052367.1 | 4 | 3921 | BAC34959.1 |

Genomic Map