Assay Details
Target Gene Details
Entrez Gene ID: | 83398 |
Gene Name: | N-deacetylase/N-sulfotransferase (heparan glucosaminyl) 3 |
Gene Aliases: |
4921531K01Rik, 4930511P15Rik, N-HSST 3, NDST-3 |
Location: |
Chr.3:123526166-123690852 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndst3 | NM_031186.3 | 1 | 792 | NP_112463.2 |
| NR_121616.1 | 2 | 940 | ||
| AF221095.1 | 1 | 790 | AAG34793.1 | |
| AK141945.1 | 2 | 941 | BAE24894.1 | |
| BC079622.1 | 1 | 707 | AAH79622.1 | |
| BC112404.1 | 1 | 703 |
More Information
Set Membership: |
|

Genomic Map