Assay Details
Target Gene Details
Entrez Gene ID: | 83398 |
Gene Name: | N-deacetylase/N-sulfotransferase (heparan glucosaminyl) 3 |
Gene Aliases: |
4921531K01Rik, 4930511P15Rik, N-HSST 3, NDST-3 |
Location: |
Chr.3:123526166-123690852 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndst3 | NM_001293682.1 | 12 | 2013 | NP_001280611.1 |
| NM_031186.3 | 13 | 3126 | NP_112463.2 | |
| NR_121616.1 | 14 | 3230 | ||
| AK141945.1 | 14 | 3231 | BAE24894.1 | |
| BC079622.1 | 13 | 2997 | AAH79622.1 |
More Information
Set Membership: |
|

Genomic Map