Assay Details
Target Gene Details
Entrez Gene ID: | 56737 |
Gene Name: | asparagine-linked glycosylation 2 (alpha-1,3-mannosyltransferase) |
Gene Aliases: |
1110018A23Rik, 1300013N08Rik, ALPG2, CDGIi, MNCb-5081 |
Location: |
Chr.4:47469833-47474374 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Alg2 | NM_019998.3 | 1 | 72 | NP_064382.3 |
| AB041604.1 | BAA95087.1 | |||
| AB161357.1 | BAD11906.1 | |||
| AK004997.2 | BAB23731.1 | |||
| AK010673.1 | BAB27106.1 | |||
| BC051951.1 | AAH51951.1 | |||
| BC052411.1 | ||||
| BY129752.1 | 1 | 73 |
More Information
Set Membership: |
|

Genomic Map