Assay Details
Target Gene Details
Entrez Gene ID: | 227644 |
Gene Name: | small nuclear RNA activating complex, polypeptide 4 |
Gene Aliases: |
5730436L13Rik |
Location: |
Chr.2:26362765-26381072 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Snapc4 | NM_001290419.1 | 22 | 2927 | NP_001277348.1 |
| NM_172339.4 | 21 | 2808 | NP_758842.1 | |
| AK077522.1 | 21 | 2775 | BAC36843.1 | |
| BC044754.2 | 6 | 1201 | AAH44754.1 | |
| BC057031.1 | 22 | 2893 | AAH57031.1 |

Genomic Map