Assay Details
Target Gene Details
Entrez Gene ID: | 27223 |
Gene Name: | transformation related protein 53 binding protein 1 |
Gene Aliases: |
53BP1, Tp53bp1, m53BP1, p53BP1 |
Location: |
Chr.2:121194835-121290320 on Build GRCm38 |
Assay Gene Location: | Within Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trp53bp1 | NM_001290830.1 | 27 | 7382 | NP_001277759.1 |
| NM_013735.4 | 28 | 7532 | NP_038763.3 | |
| AK036324.1 | 9 | 3337 | BAC29383.1 | |
| AK045416.1 | 1 | 1583 | ||
| BE943701.1 | 1 | 212 |
Target Gene Details
Entrez Gene ID: | 51885 |
Gene Name: | tubulin, gamma complex associated protein 4 |
Gene Aliases: |
4932441P04Rik, 76p, D2Ertd435e, GCP-4 |
Location: |
Chr.2:121170454-121198770 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tubgcp4 | NM_001290824.1 | 18 | 2407 | NP_001277753.1 |
| NM_153387.3 | 18 | 2410 | NP_700436.1 | |
| NR_077245.2 | ||||
| NR_110985.1 | ||||
| AK034694.1 | 9 | 1938 | ||
| AK038921.1 | 18 | 2366 | BAC30170.1 | |
| AK045305.1 | 18 | 2303 | BAC32303.1 | |
| AK046132.1 | BAC32610.1 | |||
| AK050250.1 | 18 | 2408 | BAC34146.1 | |
| AK152984.1 | BAE31633.1 | |||
| AK153171.1 | BAE31776.1 | |||
| AK161774.1 | 9 | 1206 | BAE36567.1 | |
| AK177297.1 | 2 | 243 | ||
| AK209701.1 | 2 | 243 | ||
| AK213777.1 | 2 | 243 | ||
| BC029106.1 | 18 | 2374 | AAH29106.1 |
More Information
Set Membership: |
|

Genomic Map