Assay Details
Target Gene Details
Entrez Gene ID: | 56543 |
Gene Name: | potassium voltage-gated channel, Shal-related family, member 3 |
Gene Aliases: |
AW045978, Kncd3, Kv4.3 |
Location: |
Chr.3:105452093-105674002 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnd3 | NM_001039347.1 | NP_001034436.1 | ||
| AK033962.1 | BAC28529.1 | |||
| AK147574.1 | BAE28003.1 | |||
| BY283095.1 |
Target Gene Details
Entrez Gene ID: | 78635 |
Gene Name: | potassium voltage-gated channel, Shal-related family, member 3, opposite strand |
Gene Aliases: |
1700095B22Rik |
Location: |
Chr.3:105448190-105452955 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnd3os | NR_040759.1 | 1 | 180 | |
| AK131816.1 | 1 | 180 | BAE20817.1 |
More Information
Set Membership: |
|

Genomic Map