Assay Details
Target Gene Details
Entrez Gene ID: | 66772 |
Gene Name: | ankyrin repeat and SOCS box-containing 17 |
Gene Aliases: |
4933436O18Rik |
Location: |
Chr.3:153844247-153853615 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Asb17 | NM_025758.4 | 1 | 84 | NP_080034.2 |
| AF425641.1 | 1 | 84 | AAL60518.1 | |
| AK016273.1 | 1 | 84 | ||
| AK017090.1 | 1 | 66 | ||
| BC049572.1 | 1 | 89 | AAH49572.1 |

Genomic Map