Assay Details
Target Gene Details
Entrez Gene ID: | 12870 |
Gene Name: | ceruloplasmin |
Gene Aliases: |
D3Ertd555e |
Location: |
Chr.3:19956987-20009750 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cp | NM_001276248.1 | NP_001263177.1 | ||
| NM_001276250.1 | NP_001263179.1 | |||
| NM_007752.3 | 19 | 3456 | NP_031778.2 | |
| AK014835.1 | ||||
| AK043248.1 | ||||
| AK080701.1 | 11 | 1552 | BAC37984.1 | |
| AK086999.1 | BAC39781.1 | |||
| BC062957.1 | 19 | 3456 | AAH62957.1 | |
| BE689746.1 | 1 | 48 | ||
| DQ336396.1 | 19 | 3454 | ABC68391.1 | |
| U49430.1 | AAB07996.1 |
More Information
Set Membership: |
|

Genomic Map