Assay Details
Target Gene Details
Entrez Gene ID: | 16196 |
Gene Name: | interleukin 7 |
Gene Aliases: |
A630026I06Rik, Il-7, hlb368 |
Location: |
Chr.3:7572028-7613427 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il7 | NM_008371.4 | 1 | 88 | NP_032397.1 |
| AK041307.1 | 1 | 89 | BAC30899.1 | |
| AK042009.1 | 1 | 403 | ||
| AK052452.1 | 1 | 422 | ||
| AK145184.1 | 1 | 384 | BAE26279.1 | |
| BC110553.2 | AAI10554.1 | |||
| X07962.1 | 1 | 388 | CAA30779.1 |

Genomic Map