Assay Details
Target Gene Details
Entrez Gene ID: | 55980 |
Gene Name: | inositol (myo)-1(or 4)-monophosphatase 1 |
Gene Aliases: |
2610002K09Rik, 2900059K10Rik, AI325909 |
Location: |
Chr.3:10313540-10331439 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Impa1 | NM_018864.5 | 9 | 2014 | NP_061352.2 |
| AF042730.1 | 9 | 1959 | AAB97469.1 | |
| AK011770.1 | 9 | 1952 | BAB27831.1 | |
| AK078406.1 | 9 | 1963 | BAC37259.1 | |
| AK083238.1 | 9 | 1948 | BAC38823.1 | |
| AK147778.1 | 9 | 1974 | BAE28132.1 | |
| AK152774.1 | BAE31486.1 | |||
| AK153457.1 | BAE32009.1 | |||
| AK160203.1 | 9 | 1952 | BAE35691.1 | |
| AK160467.1 | 9 | 1946 | BAE35806.1 | |
| AK165977.1 | BAE38496.1 | |||
| AK168457.1 | BAE40356.1 | |||
| BC049080.1 | 8 | 2756 | AAH49080.1 | |
| BC055722.1 | 8 | 1909 | AAH55722.1 |
More Information
Set Membership: |
|

Genomic Map