Assay Details
Target Gene Details
Entrez Gene ID: | 360213 |
Gene Name: | tripartite motif-containing 46 |
Gene Aliases: |
TRIFIC |
Location: |
Chr.3:89234169-89246154 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trim46 | NM_001039466.1 | 9 | 2543 | NP_001034555.1 |
| NM_183037.2 | 10 | 2944 | NP_898858.1 | |
| NR_121683.1 | 11 | 3112 | ||
| AK132368.1 | 9 | 2513 | ||
| AK170904.1 | 9 | 2544 | BAE42105.1 | |
| BU701363.1 |
More Information
Set Membership: |
|

Genomic Map