Assay Details
Target Gene Details
Entrez Gene ID: | 69073 |
Gene Name: | keratinocyte differentiation factor 1 |
Gene Aliases: |
1810019J16Rik |
Location: |
Chr.4:133518963-133530790 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kdf1 | NM_001083916.1 | 4 | 1624 | NP_001077385.1 |
| NM_133707.2 | 4 | 1565 | NP_598468.2 | |
| AK007551.1 | 3 | 1490 | BAC25177.1 | |
| AK158383.1 | 3 | 2092 | ||
| BC006890.1 | 4 | 1493 | AAH06890.1 | |
| BF318332.1 | 1 | 181 | ||
| BG069889.2 | 1 | 170 |

Genomic Map