Assay Details
Target Gene Details
Entrez Gene ID: | 140577 |
Gene Name: | ankyrin repeat domain 6 |
Gene Aliases: |
- |
Location: |
Chr.4:32804027-32950884 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankrd6 | NM_001012450.1 | 17 | 2570 | NP_001012453.1 |
| NM_001012451.1 | 16 | 2281 | NP_001012454.1 | |
| NM_080471.3 | 16 | 2410 | NP_536719.2 | |
| AK133551.1 | 15 | 2297 | ||
| AK173070.1 | 16 | 2321 | BAD32348.1 | |
| AY026320.2 | 15 | 2202 | AAK15806.2 | |
| BC065177.1 | 17 | 2520 | AAH65177.1 |

Genomic Map