Assay Details
Target Gene Details
Entrez Gene ID: | 140577 |
Gene Name: | ankyrin repeat domain 6 |
Gene Aliases: |
- |
Location: |
Chr.4:32804027-32950884 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankrd6 | NM_001012450.1 | 3 | 598 | NP_001012453.1 |
| NM_001012451.1 | 2 | 309 | NP_001012454.1 | |
| NM_080471.3 | 2 | 438 | NP_536719.2 | |
| AK083265.1 | 1 | 1827 | ||
| AK133551.1 | 1 | 325 | ||
| AK135024.1 | 2 | 293 | ||
| AK173070.1 | 2 | 349 | BAD32348.1 | |
| AY026320.2 | 1 | 230 | AAK15806.2 | |
| BC065177.1 | 3 | 548 | AAH65177.1 | |
| CF531081.1 | 2 | 438 | ||
| CF533090.1 | 2 | 324 |

Genomic Map