Assay Details
Target Gene Details
Entrez Gene ID: | 101113 |
Gene Name: | sorting nexin family member 21 |
Gene Aliases: |
5730407K14Rik, AI481716 |
Location: |
Chr.2:164785762-164793810 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Snx21 | NM_133924.3 | 1 | 249 | NP_598685.2 |
| AK017516.1 | 1 | 254 | BAB30785.1 | |
| AK141663.1 | 1 | 250 | BAE24791.1 | |
| AK168473.1 | 1 | 236 | ||
| AK183626.1 | ||||
| BC117542.1 | 1 | 458 |
More Information
Set Membership: |
|

Genomic Map