Assay Details
Target Gene Details
Entrez Gene ID: | 20667 |
Gene Name: | SRY (sex determining region Y)-box 12 |
Gene Aliases: |
2010205A02Rik |
Location: |
Chr.2:152393611-152398046 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sox12 | NM_011438.2 | 1 | 3701 | NP_035568.1 |
| AK081438.1 | 1 | 823 |
More Information
Set Membership: |
|

Genomic Map