Assay Details
Target Gene Details
Entrez Gene ID: | 66421 |
Gene Name: | RIKEN cDNA 2410004B18 gene |
Gene Aliases: |
4930572C24Rik |
Location: |
Chr.3:145938003-145944829 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2410004B18Rik | NM_025555.4 | 3 | 580 | NP_079831.1 |
| AK010378.1 | 3 | 580 | BAB26896.1 | |
| AK019404.1 | 3 | 576 | BAB31704.1 | |
| AK019806.1 | 2 | 224 | ||
| AK077565.1 | 4 | 698 | ||
| BC115465.2 | 3 | 579 |

Genomic Map