Assay Details
Target Gene Details
Entrez Gene ID: | 241877 |
Gene Name: | solute carrier family 10 (sodium/bile acid cotransporter family), member 5 |
Gene Aliases: |
Gm405, mP5 |
Location: |
Chr.3:10331734-10335656 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc10a5 | NM_001010834.2 | 1 | 1970 | NP_001010834.1 |
| AK143900.1 | 1 | 1971 | BAE25589.1 | |
| BC141377.1 | 1 | 1947 |
More Information
Set Membership: |
|

Genomic Map